Learn More
Electron Microscopy Sciences Molding Cup Tray, 5 mm height
SDP
Catalog No. 5024981
Encompass_Preferred
These polyethylene trays are ideal for casting specimen blocks embedded with JB-4 or equivalents and are used with our block holders and chucks shown above.
Size: 6 x 8 mm
Height: 5 mm
Catalog No. 50-249-81 Supplier Electron Microscopy Sciences Supplier No. 7017620
These polyethylene trays are ideal for casting specimen blocks embedded with JB-4 or equivalents and are used with our block holders and chucks shown above.
Size: 6 x 8 mm
Height: 5 mm