Learn More
Electron Microscopy Sciences Polyethylene trays are ideal for casting specimen blocks embedded with JB-4 or equivalents. Used with our block holders and chucks.
SDP
Catalog No. NC3306529
Encompass_Preferred
These polyethylene trays are ideal for casting specimen blocks embedded with JB-4 or equivalents and are used with our block holders and chucks shown above.
Catalog No. NC3306529 Supplier Electron Microscopy Sciences Supplier No. 7017630
These polyethylene trays are ideal for casting specimen blocks embedded with JB-4 or equivalents and are used with our block holders and chucks shown above.