Learn More
MSC Lysol Bleach Mold Mildew Remover, 32 oz Unscented Spray, 12/Carton
Catalog No. 502870159
Encompass
This product is a liquid mold and mildew remover with bleach, formulated for effective cleaning and disinfection in various environments. It comes in a 32 oz spray bottle.
  • Unscented liquid disinfectant and mold and mildew cleaner
  • Primary chemical is Sodium Hypochlorite
  • Designed for use as a bathroom cleaner
  • Suitable for ceramic material applications
  • Effective on surfaces such as sinks, vinyl shower curtains, bathtubs, counters, and glazed ceramic tile
  • Acts as a disinfectant and is specifically formulated for mold and mildew cleaning
Catalog No. 50-287-0159 Supplier MSC Supplier No. 91511337
This product is a liquid mold and mildew remover with bleach, formulated for effective cleaning and disinfection in various environments. It comes in a 32 oz spray bottle.
  • Unscented liquid disinfectant and mold and mildew cleaner
  • Primary chemical is Sodium Hypochlorite
  • Designed for use as a bathroom cleaner
  • Suitable for ceramic material applications
  • Effective on surfaces such as sinks, vinyl shower curtains, bathtubs, counters, and glazed ceramic tile
  • Acts as a disinfectant and is specifically formulated for mold and mildew cleaning