Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Mole Set
GREENER_CHOICESCI_ED

Catalog No. S139418 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139418 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139418 Supplier Supertek™ Supplier No. PH10037
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The four specimens in this Supertek™ Mole Set helps the students to explain concept of mole. This Mole set contains four element specimens: Copper, Iron, Zinc, and Aluminum. Each sample contains one mole 6.02 X 10 atoms of the element.

  • Set helps to explain concept of mole.
  • 4 element specimens
  • 1 - Copper sample
  • 1 - Iron sample
  • 1 - Zinc sample
  • 1 - Aluminum sample
  • Each sample contains one mole: 6.02 x 10 atoms of the element.

Specifications

Grade 6+
Format Set
Languages English
Subject Chemistry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.