Learn More
Erlab™ Molecode-F Formaldehyde Detector Alarm For Captair Line
GREENER_CHOICE
Catalog No. 08444097 Shop All Erlab Products
Change view
Click to view available options
Filter Type:
Carbon with Formaldehyde Sensor
Catalog No. Filter Type
08-444-097 Carbon with Formaldehyde Sensor
1 options
This item is not returnable. View return policy
Catalog No. 08-444-097 Supplier Erlab™ Supplier No. MOLECODEF
Add to Cart
Add to Cart
This item is not returnable. View return policy
Formaldehyde detector alarm for smart ductless fume hoods, weighing stations, and smart storage cabinets.
Detection sensor used to monitor post-primary filter values.
  • Audible and visual alarms if detection occurs
  • For use with Captair Ductless Filter Fume Hoods, CaptairStore Filtered Storage Cabinets, CaptairFlow Laminar Flow Hoods, and CaptairBio PCR Workstations
Specifications
Product Type Detection Alarm
Coating Epoxy
Filter Type Carbon with Formaldehyde Sensor
Lighting Type LED
Material Epoxy
Product Line Captair Flex
For Use With (Equipment) Captair, CaptairBio, CaptairFlow, CaptairStore
Includes Formaldehyde Sensor
IP Rating None
Voltage 0/1 V
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.