Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erlab™ Molecode-S Solvent Detector Alarm For Captair Line
GREENER_CHOICE

Catalog No. 08444095 Shop All Erlab Products
Change view
Click to view available options
For Use With (Equipment):
Captair, CaptairBio, CaptairFlow, CaptairStore
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
08-444-095 Captair, CaptairBio, CaptairFlow, CaptairStore
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 08-444-095 Supplier Erlab™ Supplier No. MOLECODES
Add to Cart
Add to Cart
This item is not returnable. View return policy

Solvent detector alarm for smart ductless fume hoods, weighing stations, and smart storage cabinets.

Detection sensor used to monitor post-primary filter values.

  • Audible and visual alarms if detection occurs
  • For use with Captair Ductless Filter Fume Hoods, CaptairStore Filtered Storage Cabinets, CaptairFlow Laminar Flow Hoods, and CaptairBio PCR Workstations

Specifications

For Use With (Equipment) Captair, CaptairBio, CaptairFlow, CaptairStore
Includes Solvent Sensor
Product Type Detection Alarm
Voltage 0/1 V
Material Metal Oxide
For Use With (Application) monitoring post-primary filter values

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.