Learn More
IBI Scientific™ Molecular Biology Grade Agarose
Catalog No. IB70045 Shop All Stovall Life Science Inc. Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IB70045 1 kg
IB70041 250 g
IB70035 25 g
IB70042 500 g
IB70040 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. IB70045 Supplier IBI Scientific™ Supplier No. IB70045
Add to Cart
Add to Cart
Molecular biology certified for use in standard electrophoresis procedures including nucleic acid, analytical, and preparative electrophoresis and blotting. IBI Scientific™ Molecular Biology Grade Agarose is highly refined and purified with strict testing parameters to insure lot-to-lot consistency.
  • Mechanical resistance
  • Low absorption of staining reagents
  • Transparency for high visibility
  • Thermal stability due to high hysteresis
  • Lot-to-lot consistency
  • DNase- and RNase-Free
Specifications
For Use With (Application) Electrophoresis
Quantity 1 kg
Product Type Agarose Gel