Learn More
IBI SCIENTIFIC 3P Molecular Biology Grade Water, 125 mL
Catalog No. NC0753501
Encompass_Preferred
Molecular Biology Grade Water 125 ml Certified nuclease-free and pyrogen-free 0.1 m sterile-filtered into certified-clean polycarbonate bottles Steam-sterilized (autoclaved twice) 18.2 Megohm deionized Available in multiple sizes; sold individually and in multi-bottle cases Applications: PCR, RT-PCR, restriction enzyme assays, modifying enzyme assays, transfection, cloning, transformation, and all general molecular biology lab procedures
Catalog No. NC0753501 Supplier IBI SCIENTIFIC 3P Supplier No. IB42100
Molecular Biology Grade Water 125 ml Certified nuclease-free and pyrogen-free 0.1 m sterile-filtered into certified-clean polycarbonate bottles Steam-sterilized (autoclaved twice) 18.2 Megohm deionized Available in multiple sizes; sold individually and in multi-bottle cases Applications: PCR, RT-PCR, restriction enzyme assays, modifying enzyme assays, transfection, cloning, transformation, and all general molecular biology lab procedures