Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Molecular Biology Reagents
BIOPHARMAGSA_VA

Catalog No. MT46031CM Shop All Corning Products
Change view
Click to view available options
Chemical Name or Material:
Saline Sodium Citrate Buffer
TAE Buffer
Tris Borate EDTA Buffer
Tris Hydrochloride Buffer
pH:
7 ±0.1
7.5 ±0.1
8 ±0.1
8.3 ±0.1
8.4 ±0.1
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material pH
MT46031CM Tris Hydrochloride Buffer 8 ±0.1
MT46010CM TAE Buffer 8.3 ±0.1
MT46030CM Tris Hydrochloride Buffer 7.5 ±0.1
MT46011CM Tris Borate EDTA Buffer 8.4 ±0.1
MT46020CM Saline Sodium Citrate Buffer 7 ±0.1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. MT46031CM Supplier Corning™ Supplier No. 46031CM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in molecular biology, nucleic acid purification applications

  • Certificate of Analysis available
  • Product can be diluted to obtain the desired concentration

Order Info

Shipped at ambient temperature

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris Hydrochloride Buffer
CAS 1185-53-1, 7732-18-5
Concentration 1 M
For Use With (Application) Molecular Biology
pH 8 ±0.1
Physical Form Liquid
Quantity 6 x 1 L
Solution Type Buffer
Sterility Sterile
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.