Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Molecular BioProducts™ HLT Ergo-F Pipet Tips
GREENER_CHOICE

Catalog No. 21403474 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-403-474 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-403-474 Supplier Thermo Scientific™ Supplier No. 3812
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Finnpipette Focus and Digital Pipetters

  • Ergonomic tips reduce strains and injury
  • Secure and consistent seals
  • Less ejection force
  • Also available in low retention polymer

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Sterility Non-sterile
Format Rack
For Use With (Equipment) Thermo Scientific Finnpipette Focus, Digital Pipette
Product Line Molecular BioProducts HLT Ergo-F
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.