Learn More
Fluotics Hamilton Star and Vantage Compatible P50, filter/sterile
Catalog No. 5000435183
Encompass_Preferred
Hamilton Star and Vantage Compatible P50, filter/sterile, Working Volume: 40uL, Tip Color: Clear, Single Stack, 384 Multi-Probe Head CO-RE, Case of 15,360 tips (40 NTR style stacks; 384/stack4
Catalog No. 50-004-35183 Supplier Fluotics Supplier No. HAM50.F
Hamilton Star and Vantage Compatible P50, filter/sterile, Working Volume: 40uL, Tip Color: Clear, Single Stack, 384 Multi-Probe Head CO-RE, Case of 15,360 tips (40 NTR style stacks; 384/stack4