Learn More
Molecular Dimensions Molecular Dimensions Precipitant Mix 3, Morpheus, 250 mL
SDP
Catalog No. NC3974965
Encompass
This precipitant mix is a key component of the Morpheus protein crystallization screen, specifically formulated for 3D protein crystallization research and development. It is designed to provide cryo-protection for samples, thanks to its glycerol content.
  • Contains 40% v/v glycerol.
  • Contains 20% w/v PEG 4000.
  • Supplied in a 250 mL volume.
  • Part of a screen designed for 3D protein crystallization.
  • Includes a cryo-agent for cryo-protected conditions.
Catalog No. NC3974965 Supplier Molecular Dimensions Supplier No. MD225083
This precipitant mix is a key component of the Morpheus protein crystallization screen, specifically formulated for 3D protein crystallization research and development. It is designed to provide cryo-protection for samples, thanks to its glycerol content.
  • Contains 40% v/v glycerol.
  • Contains 20% w/v PEG 4000.
  • Supplied in a 250 mL volume.
  • Part of a screen designed for 3D protein crystallization.
  • Includes a cryo-agent for cryo-protected conditions.