Learn More
Research Products International Corp Water, Molecular Biology Grade, DNase and RNase Free, 1 Gallon
SDP
Catalog No. 50212868
Encompass_Preferred
Certified RNase and DNase free for use in molecular biology applications. Ideal for making reagents, rinsing glass and plastic ware, enzymatic reactions and more. Toxic agents such as DEPC are not used in purification procedures.
  • Endotoxin Free
  • RNase Free
  • DNase Free
*Tested to < 0.001 EU/ML
Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water.
Catalog No. 50-212-868 Supplier Research Products International Corp Supplier No. 248700
Add to Cart
Add to Cart
Certified RNase and DNase free for use in molecular biology applications. Ideal for making reagents, rinsing glass and plastic ware, enzymatic reactions and more. Toxic agents such as DEPC are not used in purification procedures.
  • Endotoxin Free
  • RNase Free
  • DNase Free
*Tested to < 0.001 EU/ML
Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water.