Learn More
ATCC Molecular Grade Water 1L
Catalog No. 502382355
Encompass
Molecular grade water is DNase, RNase, and Protease free and can be used in all molecular biology procedures that require water, including PCR.
Catalog No. 50-238-2355 Supplier ATCC Supplier No. 602450
Molecular grade water is DNase, RNase, and Protease free and can be used in all molecular biology procedures that require water, including PCR.