Learn More
Invitrogen™ Albumin from Bovine Serum (BSA), Alexa Fluor™ 555 conjugate
Description
This albumin from bovine serum (BSA), Alexa Fluor 555 conjugate is a bright and photostable conjugate. Can be used for applications that include studying the rate of endocytosis and exocytosis and following intracellular protein processing.
Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 5 mg |
| Conjugate | Alexa Fluor 555 |
| Species | Bovine |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Label Type | Alexa Fluor Dyes |
| Shipping Condition | Room Temperature |
| Form | Lyophilized |
| Protein Subtype | Albumin Proteins |
| Product Line | Alexa Fluor |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.