Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Albumin from Bovine Serum (BSA), Tetramethylrhodamine conjugate

Catalog No. A23016
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23016 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A23016 Supplier Invitrogen™ Supplier No. A23016
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Suited for applications in which well-defined molecular weight tracers are needed

  • Applications include studying rate of endocytosis and exocytosis and following intracellular protein processing

Cell Analysis, Cell Tracing and Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Species Bovine
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Classic Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Albumin Proteins

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.