Learn More
Invitrogen™ Isolectin GS-IB4 From Griffonia simplicifolia, Alexa Fluor™ 594 Conjugate
Description
Isolectin GS-IB4 is a 114,000-dalton glycoprotein that is part of a family of five tetrameric type I isolectins (IA4, IA3B, IA2B2, IAB3, and IB4. The A subunit prefers N-acetyl-D-galactosamine end groups while the B subunit is selective for terminal a-D-galactosyl residues. The red fluorescent Alexa Fluor 594 isolectin GS-IB4 conjugate can be used in fluorescence microscopy.
Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Microbial Tracking
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 500 μg |
| Source | Africa Shrub legume |
| Conjugate | Alexa Fluor 594 |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Label Type | Alexa Fluor Dyes |
| Shipping Condition | Room Temperature |
| Form | Lyophilized |
| Protein Subtype | Lectins |
| Product Line | Alexa Fluor |
Frequently Asked Questions (FAQs)
Factors to consider are size of tracer, method of delivery (injection, direct application to tissue, etc.), and if the tracer needs to be fixable. Here are some links to details about the various classes of neuronal tracers we offer and how to choose between them:
Neuronal Tracing (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing.html)
Choosing a Tracer (https://www.thermofisher.com/us/en/home/references/molecular-probes-the-handbook/fluorescent-tracers-of-cell-morphology-and-fluid-flow/choosing-a-tracer.html)
Imaging Analysis (http://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/bioprobes-50-journal.pdf)
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.