Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cholera Toxin Subunit B (Recombinant) Conjugate

Catalog No. C34776
Encompass_Preferred
Change view
Click to view available options
Conjugate:
Alexa Fluor 488
Alexa Fluor 555
Alexa Fluor 594
Alexa Fluor 647
Biotin
HRP (Horseradish Peroxidase)
Quantity:
100 μg
500 μg
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate Quantity
C34776 Alexa Fluor 555 100 μg
C22841 Alexa Fluor 488 500 μg
C22843 Alexa Fluor 555 500 μg
C34777 Alexa Fluor 594 100 μg
C22842 Alexa Fluor 594 500 μg
C34778 Alexa Fluor 647 100 μg
C34779 Biotin 100 μg
C34780 HRP (Horseradish Peroxidase) 100 μg
C34775 Alexa Fluor 488 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. C34776 Supplier Invitrogen™ Supplier No. C34776
  • $360.00
    $341.65 / Each of 1

    Save $18.35

    5% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant cholera toxin B subunit (CT-B) conjugates bind with high affinity to GM1 ganglioside and support neuronal tracing, GM1 labeling, membrane trafficking, and membrane organization studies. Choose from Alexa Fluor™ 488, 555, 594, and 647 fluorescent conjugates, Biotin-XX, or horseradish peroxidase (HRP) to match your detection workflow.

Recombinant cholera toxin B conjugates for GM1 labeling and neuronal tracing

Cholera toxin B subunit (CT-B) binds with high affinity to GM1 ganglioside on the cell surface and is widely used for neuronal tracing, GM1 labeling, endocytosis, and membrane trafficking studies. At neutral pH, CT-B forms a pentamer with multiple GM1-binding sites, enabling multivalent interactions with GM1-containing membranes.

These conjugates are made using recombinant cholera toxin B subunit without the enzymatically active A subunit of native cholera toxin.

Why use recombinant cholera toxin B conjugates?

  • Label GM1 ganglioside: Use the high-affinity interaction between CT-B and GM1 to visualize and detect GM1-associated membrane structures.
  • Trace neuronal projections: Use CT-B for well-established retrograde labeling of neuronal pathways.
  • Study membrane trafficking: Follow CT-B/GM1 internalization and retrograde trafficking through endosomal pathways.
  • Investigate membrane organization: Label GM1 and examine GM1-associated membrane domains in lipid raft-related studies.
  • Use recombinant B subunit: Study GM1 binding and trafficking with CT-B lacking the enzymatically active A subunit of native cholera toxin.
  • Choose multiple detection formats: Select fluorescent Alexa Fluor™ conjugates, Biotin-XX, or HRP to match fluorescence, affinity-based, or enzyme-based detection workflows.

Choose a CT-B conjugate for your detection workflow

Select from Alexa Fluor™ 488, 555, 594, and 647 CT-B conjugates for fluorescence imaging, Biotin-XX for avidin- or streptavidin-based detection, or HRP for enzyme-based detection.

Why choose from multiple conjugates?

  • Match your imaging system: Select an Alexa Fluor™ conjugate compatible with available excitation sources, emission filters, and detectors.
  • Support multicolor imaging: Choose spectrally distinct Alexa Fluor™ dyes for use with other fluorescent probes, antibodies, and cellular markers.
  • Enable affinity-based detection: Use Biotin-XX conjugates with avidin- or streptavidin-based detection workflows.
  • Enable enzyme-based detection: Use HRP conjugates for chromogenic or chemiluminescent detection workflows.
  • Use one GM1-binding probe across workflows: Apply recombinant CT-B with different labels while maintaining the same GM1-binding biological probe.

Applications of cholera toxin B conjugates

Cholera toxin B conjugates are versatile probes for neuronal tracing, GM1 ganglioside labeling, endocytosis, and membrane trafficking studies.

Use CT-B conjugates to:

  • Perform neuronal tracing, including widely used retrograde labeling of neuronal projections
  • Label GM1 ganglioside on cell membranes
  • Visualize GM1-associated membrane domains in lipid raft-related studies
  • Study CT-B/GM1 internalization and retrograde trafficking through endosomal pathways
  • Perform fluorescence imaging with Alexa Fluor™ 488, 555, 594, or 647 conjugates
  • Perform avidin- or streptavidin-based detection with Biotin-XX conjugates
  • Perform enzyme-based detection with HRP conjugates

For Research Use Only. Not for use in diagnostic procedures.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 μg
Conjugate Alexa Fluor 555
Recombinant Recombinant
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Protein Form Recombinant
Form Lyophilized
Protein Subtype Cholera Toxin
Product Line Alexa Fluor
I injected a fluorescent tracer, but cannot detect it after tissue is fixed and sectioned. What am I doing wrong?

Confirm that the tracer you are using crosslinks to proteins or has a primary amine for fixation-either a hydrazide, lysine fixable dextran, or a protein conjugate.
Use aldehyde-based fixatives to cross link the amines on the tracer.
Inject a larger amount or higher concentration of the tracer. Tracers are generally injected at 1-20% concentrations (10 mg/mL or higher).
Confirm that you are using the correct fluorescent filter for detection. You can perform a spot test by pipetting a small amount of the undiluted stock solution of the tracer onto a slide, then view under the filter you are using on your microscope. This will confirm if the tracer fluorescence can be detected and the fluorescent microscope filter is working properly.
Review tissue fixation and handling procedures to confirm if any reagents or processing procedures could be affecting the tracer.

Do you have a tracer that will only transport retrograde?

Wheat germ agglutinin and cholera toxin conjugates have been used for retrograde tracing. They may have some anterograde tracing in some applications. A selection guide can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing/protein-conjugates.html).

How do I know which tracer to choose for my experiment?

Factors to consider are size of tracer, method of delivery (injection, direct application to tissue, etc.), and if the tracer needs to be fixable. Here are some links to details about the various classes of neuronal tracers we offer and how to choose between them:

Neuronal Tracing (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing.html)
Choosing a Tracer (https://www.thermofisher.com/us/en/home/references/molecular-probes-the-handbook/fluorescent-tracers-of-cell-morphology-and-fluid-flow/choosing-a-tracer.html)
Imaging Analysis (http://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/bioprobes-50-journal.pdf)

What products do you have for neuronal tracing?

Please check out this web page (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing.html) for details.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.