Learn More
Invitrogen™ Cholera Toxin Subunit B (Recombinant), Horseradish Peroxidase Conjugate
Description
- Cholera toxin B subunit (CT-B) attaches to cells by binding to ganglioside GM1, making it powerful tool for retrograde labeling of neurons
- This tracer has been used in variety of applications, including tracing of rat forebrain afferents, projections of the parabrachial region, and neurons of the urinary bladder wall
- When used in neuronal tracing applications, CT-B is typically introduced by pressure injection or by iontophoretic injection into neural tissue
- Lyophilized product can be dissolved in buffer (e.g., PBS) for use
- More recently, researchers have found that CT-B can be used as marker for lipid rafts, which are membrane microdomains enriched in cholesterol and sphingolipids thought to be important in cell signaling
- For lipid raft staining, cells are first incubated with fluorescent CT-B, then an anti-CT-B antibody is added to crosslink the CT-B in the lipid rafts into distinct patches on the plasma membrane
- These patches are easily visualized by fluorescence microscopy
- Label (Ex/Em): Horseradish peroxidase (HRP)
- At neutral pH, the 11.4 kDa B subunit exists as a 57 kDa pentamer
Cell Analysis, Cell Structure, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Lipid Rafts, Microbial Tracking, Neuronal Tracing
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Quantity | 100 μg |
| Conjugate | HRP (Horseradish Peroxidase) |
| Recombinant | Recombinant |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Label Type | Other Label(s) or Dye(s) |
| Shipping Condition | Room Temperature |
| Protein Form | Recombinant |
| Form | Lyophilized |
| Protein Subtype | Cholera Toxin |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.