Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CholEsteryl 4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoate (CholEsteryl BODIPY™ FL C12)

Catalog No. C3927MP
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3927MP 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C3927MP Supplier Invitrogen™ Supplier No. C3927MP
  • $310.00
    $254.20 / Each of 1

    Save $55.80

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Can be used as general nonexchangeable membrane marker

Cholesteryl esters consist of a fatty acid esterified to the 3β-hydroxyl group of cholesterol. These very nonpolar species are the predominant lipid components of atherosclerosis plaque and low- and high-density lipoporotein (LDL and HDL) cores. The green fluorescent BODIPY™ FL C12 cholesteryl ester can be used as a tracer of cholesterol transport and receptor-mediated endocytosis of lipoproteins by fluorescence microscopy.

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cholesterol Analogs
Quantity 1 mg
Physical Form Solid
Product Line BODIPY
How should I store CholEsteryl 4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoate (CholEsteryl BODIPY FL C12) (Cat. No. C3927MP) once reconstituted?

Once reconstituted in either chloroform or alcohol, store frozen, desiccated and protected from light. The CholEsteryl BODIPY FL C12 is not sensitive to changes in temperature (freeze/thaw), but the gradual accumulation of water from condensation and from the atmosphere will promote aggregation of the reagent over time.

What is the molecular weight of CholEsteryl 4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoate (CholEsteryl BODIPY FL C12) (Cat. No. C3927MP)? Is the molecular weight based on the cholesteryl moiety together with BODIPY?

The molecular weight of the product is 786.98 g/mol. This is the molecular weight of the full molecule.

How do I reconstitute CholEsteryl 4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoate (CholEsteryl BODIPY FL C12) (Cat. No. C3927MP)?

CholEsteryl BODIPY FL C12 is soluble up to 1 mM in chloroform. The molecular weight of this product is 786.98 g/mol. Additionally, there are references to the product being soluble up to 25 mM in ethanol and up to 0.5 mg/mL in methanol with sonication.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.