Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Click-IT™ Biotin Protein Analysis Detection Kit
GREENER_CHOICE

Catalog No. C33372
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33372 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C33372 Supplier Invitrogen™ Supplier No. C33372
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Identify and characterize glycoproteins by Western blot

The Click-iT Biotin Glycoprotein Detection Kit provides the second part of a simple and robust technique to identify and characterize glycoproteins by Western blot. In step two, after the incorporation of an azide moiety into protein glycan structures with either a Click-iT metabolic labeling reagent or the Click-iT Enzymatic Labeling system, the azide-modified glycoproteins are detected via the chemoselective ligation or click reaction between an azide and an alkyne. With this technique, detection sensitivity is in the low femptomole range and biotin-labeled samples can be detected before or after probing the Western with a primary antibody.

  • Less hazardous

Cell Analysis, Click Chemistries, Glycoprotein Analysis with Click Chemistry, Labeling Chemistry, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Biotin Protein Analysis Detection Kit
Quantity 1 Kit
Content And Storage Store in freezer -5°C to -30°C.
Detection Method Biotin-based, Fluorescence
Shipping Condition Room Temperature
Product Line Click-iT
Label or Dye Biotin
Labeling Target Proteins
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.