Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Click-iT™ RNA Alexa Fluor™ 488 HCS Assay
GREENER_CHOICE

Catalog No. C10327
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10327 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C10327 Supplier Invitrogen™ Supplier No. C10327
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

The kit contains sufficient material for two 96-well plates.

Detect newly synthesized RNA or changes in RNA levels

The Click-iT RNA Alexa Fluor 488 HCS Assay enables detection of global RNA transcription temporally and spatially in cells (PNAS (2008) 105:15779-84). The ability to detect newly synthesized RNA or changes in RNA levels resulting from disease, environmental damage or drug treatments is an important aspect of toxicological profiling. Utilizing an alkyne-modified nucleoside, 5-ethynyl uridine (EU), and powerful click chemistry, newly synthesized RNA can be detected without the use of radioactivity or antibodies with a simple, two-step procedure. In step one, the alkyne-containing nucleoside is fed to cells or animals and actively incorporated into nascent RNA. The small size of the tag enables efficient incorporation of the modified nucleoside into RNA, but not into DNA. Detection utilizes the chemoselective ligation or “click” reaction between and azide and an alkyne where the modified RNA is detected with a corresponding azide-containing dye. With its dimunitive “footprint”, the Click-iT detection molecule can easily penetrate complex samples and leaves open the possibility of multiplex analyses with other probes, including antibodies for the detection of RNA-interactive proteins for deeper biological insights.

  • Less hazardous

Cell Analysis, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, Click Chemistries, DNA and RNA Purification and Analysis, High-Content Screening (HCS), Labeling Chemistry, Nascent RNA Detection and Capture with Click Chemistry, Nucleic Acid Labeling and Oligo Synthesis, RNA Labeling

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The kit contains sufficient material for 2 X 96-well plates. Store at 2-6°C, dessicate and protect from light.
Color Green
Detection Method Fluorescence
Format 96-well Plate
Label or Dye Alexa Fluor 488
Excitation/Emission 495/519
For Use With (Equipment) High Content Instrument
Product Line Alexa Fluor, Click-iT, Molecular Probes
Product Type New RNA Synthesis Assay
Quantity 1 kit
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.