Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Coverslip Mini-Rack, for 8 coverslips

Catalog No. C14784
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14784 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C14784 Supplier Invitrogen™ Supplier No. C14784
  • $192.00
    $157.44 / Each of 1

    Save $34.56

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Miniature support designed to vertically hold eight standard round or square coverslips

  • Mini-rack fits easily into standard 50mL beaker and can accommodate small stir bar beneath rack
  • Use of mini-rack eliminates necessity for repeatedly moving coverslips between solutions with forceps
  • Ideal for sequential wash steps where thorough dilution and removal of material from coverslips is critical
  • Mini-rack is low-binding, withstands strong acids and bases, is not damaged by heat and may be sterilized by variety of methods such as autoclaving, organic solvent treatment or ethylene oxide exposure
  • Disassembles easily for cleaning and storage

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Type Coverslip Mini-Rack
Volume (Metric) Working 50 mL
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Sterility Sterile
Quantity 1 each

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.