Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CultureWell™ Chambered Coverglass for Cell Culture, 16 wells per coverglass

Catalog No. C37000
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C37000 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C37000 Supplier Invitrogen™ Supplier No. C37000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for cell culture and fluorescence imaging applications

The CultureWell removable chambered coverglass provided sterile and ready to use—contains 16 wells that can each hold up to 250μL, allowing cells to be cultured in a number of different conditions on a single slide. When the cells are ready to be imaged, coverglass removal is made easy by the use of a simple tool (Cat. No. C37010, purchased separately) that separates the parts without the need for excessive force, eliminating the risk of coverglass breakage. Frosted microscope slides are also provided for mounting. A sample size containing two chambered coverglasses and the removal tool is also available (Cat. No. C37005)

Cell Analysis, Cell Culture, Cellular Imaging, IHC Sample Preparation, IHC Staining & Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunohistochemistry (IHC), Mammalian Cell Culture

TRUSTED_SUSTAINABILITY

Specifications

Type Coverglass
Wells 16
Culture Area 0.4 cm2
Surface Treatment Cell Culture
Volume (Metric) Working 250 μL
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Sterility Sterile
Quantity 1 set
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.