Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Deoxyribonuclease I, Alexa Fluor™ 594 Conjugate

Catalog No. D12372
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12372 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12372 Supplier Invitrogen™ Supplier No. D12372
Add to Cart
Add to Cart

Can be used for detection of unpolymerized actin in fixed cells

The bright red fluorescent Alexa Fluor 594 DNase I selectively binds to G-actin (globular).

Cell Analysis, Cell Structure, Cytoskeleton

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Conjugate Alexa Fluor 594
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Excitation/Emission 590⁄617
Label or Dye Alexa Fluor™ 594
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Sub Cellular Localization Actin, Cytoskeleton
Form Lyophilized
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.