Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DQ™ Green BSA - Special Packaging

Catalog No. D12050
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12050 5 x 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12050 Supplier Invitrogen™ Supplier No. D12050
  • $598.00
    $490.36 / Each of 1

    Save $107.64

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Hydrolysis of substrate yields brightly fluorescent product

DQ Green BSA is a fluorogenic substrate for proteases. A strong fluorescence quenching effect is observed when proteins are heavily labeled with BODIPY dyes. Upon hydrolysis of the DQ Green BSA to single, dye-labeled peptides by proteases, this quenching is relieved, producing brightly fluorescent products.

Biochemical Proteasome Assays, Cell Analysis, Cell Viability, Proliferation and Function, Drug Discovery and Development, Endocytosis, Exocytosis and Phagocytosis, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteasome Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Form Solid
Product Line DQ
Quantity 5 x 1 mg
Substrate Protease Substrate
Substrate Properties Protein-Based Substrate
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.