Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELF™ 97 Phosphatase Substrate (ELF™ 97 Phosphate), 0.2 μm Filtered

Catalog No. E6588
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E6588 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E6588 Supplier Invitrogen™ Supplier No. E6588
Only null left
Add to Cart
Edge
Add to Cart

For use with microarray, flow cytometer and fluorescence microscope

The ELF 97 phosphatase substrate upon hydrolysis produces a bright and photostable yellow-green fluorescent precipitate at the site of enzymatic activity. This fluorescent precipitate has several unique spectral characteristics, including an extremely large Stokes shift that makes it easily distinguishable from endogenous fluorescence.

Cell Analysis, Cell Culture, Cell Viability, Proliferation & Function, Cellular Imaging, Differentiation & Development, Drug Discovery & Development, Enzyme & Protein Activity Assays, IHC Staining & Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining & Detection, Immunohistochemistry (IHC), Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation & Analysis, Stem Cell Analysis, Stem Cell Differentiation, Stem Cell Labeling & Detection, Stem Cell Research, Target & Lead Identification & Validation

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Cell-Permeant
Color Yellow-Green
Concentration 5 mM
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Form Liquid
Product Line ELF
Quantity 1 mL
Substrate Phosphatase Substrate
Substrate Properties Chemical Substrate
Target Enzyme Phosphatases
Excitation/Emission 345⁄530
For Use With (Equipment) Fluorescence Microscope, Microarray, Flow Cytometer
Label or Dye ELF 97
Research Category Differentiation
Shipping Condition Room Temperature
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.