Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fibrinogen From Human Plasma, Alexa Fluor™ 488 Conjugate

Catalog No. F13191
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F13191 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F13191 Supplier Invitrogen™ Supplier No. F13191
Add to Cart
Edge
Add to Cart

For investigating platelet activation and subsequent fibrinogen binding

Molecular Probes fibrinogen conjugates are prepared by attaching fluorescent dye molecules to purified human fibrinogen (approximately 15 dye molecules for each fibrinogen molecule), purifying the conjugate to remove unreacted dye, and then lyophilizing for storage. Fluorescently labeled fibrinogen has proven to be a valuable tool for investigating platelet activation and subsequent fibrinogen binding. For instance, fluorescein-labeled fibrinogen has been used to detect fibrinogen bound to activated platelets by flow cytometry.

  • Label (Ex/Em): Alexa Fluor 488 (approx. 495/519nm)
  • Spectrally similar to fluorescein, but the signal is less pH-dependent and more photostable
  • Lyophilized product can be dissolved in buffer (e.g., sodium bicarbonate, pH 8.3) for use
  • Fluorescence is typically detected using fluorescence microscopy or flow cytometry

Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Conjugate Alexa Fluor 488
Species Human
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Fibrinogen
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.