Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FocalCheck™ Fluorescent Microspheres Kit, 6 μm, mounted on slides

Catalog No. F24633
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F24633 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F24633 Supplier Invitrogen™ Supplier No. F24633
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Beads are pre-mounted on slides for confocal microscope calibration

Our popular FocalCheck microspheres have been available in suspension for several years. Now, in response to customer requests, we offer FocalCheck beads pre-mounted on microscope slides. The 6 μm FocalCheck Fluorescent Microsphere Kit features mounted samples of our three best-selling color configurations: fluorescent green/orange/dark red ring stains, fluorescent green ring stain/blue throughout and fluorescent dark red ring stain/green throughout.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Calibration Type Confocal Microscope Calibration
Content And Storage Store at room temperature and protect from light.
Color Orange, Dark Red, Blue, Green
Product Type Fluorescent Microspheres Kit
Format Mounted on Slide
Product Line FocalCheck
Quantity 1 Kit
Shipping Condition Room Temperature
Diameter (Metric) 6 μm
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.