Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FocalCheck™ Microspheres, 15 μm, fluorescent green/orange/dark-red ring stains

Catalog No. F7235
Encompass_Preferred
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F7235 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F7235 Supplier Invitrogen™ Supplier No. F7235
Add to Cart
Edge
Add to Cart

Fluorescent green/orange/dark-red ring stains

The sharp ring stains exhibited by the 15μm FocalCheck fluorescent green/orange/dark-red ring stained microspheres produce a striking visual representation of instrument misalignment or other aberrations making them ideal as reference standards for confocal laser-scanning microscopy. Correct image registration is indicated when the multiple ring images are perfectly coincident in all dimensions.

Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)

TRUSTED_SUSTAINABILITY

Specifications

Calibration Type Confocal Microscope Calibration
Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Color Orange, Dark Red, Green
Product Type Microsphere
Format Suspension Beads
Product Line FocalCheck
Quantity 0.5 mL
Shipping Condition Room Temperature
Diameter (Metric) 15 μm

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.