Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Guanosine 5'-Triphosphate, BODIPY™ FL 2'-(or-3')-O-(N-(2-Aminoethyl) Urethane), Trisodium Salt (BODIPY™ FL GTP)

Catalog No. G12411
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G12411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G12411 Supplier Invitrogen™ Supplier No. G12411
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Based on interactions between the fluorophore and the purine base

BODIPY FL adenosine 5'-triphosphate (A-12410) and BODIPY FL guanosine 5'-triphosphate are composed of a BODIPY FL fluorophore attached to the 2' or 3'' position of the ribose ring via a linker. Interactions between the fluorophore and the purine base are evident from the spectroscopic properties of these nucleotide analogs. The fluorescence quantum yield of BODIPY FL GTP is only about one-tenth that of BODIPY FL ATP; however, this yield may be enhanced upon binding of BODIPY FL GTP to GTP-binding proteins.

Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Guanosine 5'-Triphosphate
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Shipping Condition Wet Ice
Concentration 5 mM
Label or Dye BODIPY™ FL
Quantity 100 μL
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.