Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Heparin, Fluorescein Conjugate

Catalog No. H7482
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H7482 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H7482 Supplier Invitrogen™ Supplier No. H7482
Add to Cart
Edge
Add to Cart

Shows heparin binding in cells and tissue

The green-fluorescent fluorescein heparin conjugate should be a useful tool for studying binding of this mucopolysaccharide in cells and tissue.

Cell Analysis, Cell Structure, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Receptors

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Conjugate Fluorescein
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Classic Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Other Proteins
How can I dissolve Heparin, Fluorescein Conjugate (Cat. No. H7482)?

The molecular weight of Heparin, Fluorescein Conjugate (Cat. No. H7482) is ~18,000 and the information can be found on the Calcium Regulation—Section 17.2 page (see Data Table).

Heparin is quite soluble in water (10mg/ml) or aqueous buffer and you can prepare stock solutions of at least 1 mM using water as the solvent. Once dissolved, the stability of the nonsterile solution is very poor, so it should either be sterile filtered (0.2 µm) or an antibacterial (e.g., 0.06-1% benzyl alcohol) should be added. Heparin in solution should not be frozen, but is stable at 2-8 degrees C.

What is size of the Heparin, Fluorescein Conjugate (Cat. No. H7482)?

Heparin is an irregular aggregate of a 3,000 MW monomers, and they are low molecular weight heparins. They are purified by dialysis with a cut off of > 10 kDa and have an average of 18kDa.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.