Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lectin GS-II From Griffonia simplicifolia, Alexa Fluor™ 488 Conjugate

Catalog No. L21415
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L21415 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L21415 Supplier Invitrogen™ Supplier No. L21415
Only null left
Add to Cart
Edge
Add to Cart

Isolated from the seeds of Griffonia simplicifolia, a tropical African legume

Lectin GS-II is a 113,000-dalton compoased of identical subunits. Each subunit contains a single binding site specific for terminal non-reducing a- or Β-linked N-acetyl-D-glucoasemine. Unlike many other lectins, GS-II does not react with any specific blood group. The green-fluorescent lectin GS-II Alexa Fluor 488 conjugate can be used in fluorescence microscopy or flow cytometry.

Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Microbial Tracking

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 μg
Source Africa Shrub legume
Conjugate Alexa Fluor 488
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Lectins
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.