Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lectin HPA From Helix pomatia (edible snail), Alexa Fluor™ 647 Conjugate

Catalog No. L32454
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L32454 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L32454 Supplier Invitrogen™ Supplier No. L32454
Add to Cart
Edge
Add to Cart

Suitable for use in cellular imaging and flow cytometry

Lectin Helix pomatia agglutinin (HPA) selectively binds to α-N-acetylgalactosamine residues and type A erythrocytes.

Cell Analysis, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Source Edible snail
Conjugate Alexa Fluor 647
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Lectins
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.