Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lectin PHA-L From Phaseolus vulgaris (red kidney bean), Alexa Fluor™ 594 Conjugate

Catalog No. L32456
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L32456 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L32456 Supplier Invitrogen™ Supplier No. L32456
Add to Cart
Edge
Add to Cart

Suitable for use in cell tracing and tracking, cellular imaging and flow cytometry

Lectin phytohemagglutinin-L (PHA-L) is strongly inhibited by N-acetylglucosamine β(1-2) mannopyranosyl residues.[ALTREF PN29369] This lectin has been shown to agglutinate leukocytes and induce mitogenic activity in T-lymphocytes[ALTREF PN29369,PN29368] and has also been used as an anterograde tracer for the study of neuronal pathways.[ALTREF PN20284]

Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Microbial Tracking

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Source Red kidney bean
Conjugate Alexa Fluor 594
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Lectins
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.