Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lectin PNA From Arachis hypogaea (peanut), Alexa Fluor™ 488 Conjugate

Catalog No. L21409
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L21409 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L21409 Supplier Invitrogen™ Supplier No. L21409
  • $230.00
    $220.65 / Each of 1

    Save $9.35

    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Specific for terminal β-galactose

Lectin PNA will agglutinate human erythrocytes, but only after neuraminidase treatment.[ALTREF PN40265]

Cell Analysis, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Flow Cytometry, Flow Cytometry Antibodies and Secondary Detection, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Microbial Tracking

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Source Peanut
Conjugate Alexa Fluor 488
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Lectins
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.