Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LI Silver (LIS) Enhancement Kit

Catalog No. L24919
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L24919 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L24919 Supplier Invitrogen™ Supplier No. L24919
Add to Cart
Edge
Add to Cart

Convenient, light-insensitive silver enhancement system for use with colloidal gold and NANOGOLD reagents

Gold particles in the presence of silver (I) ions and a reducing agent act as catalysts to reduce the silver (I) ions to metallic silver. The metallic silver is deposited onto the gold, enlarging the particles to between 30 and 100nm in diameter. Tissue or blots stained with colloidal gold are “developed” by this autometallographic procedure to give black staining that can be seen in the light microscope.

The LI Silver Enhancement Kit is prepared for Molecular Probes by Nanoprobes, Inc.

Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C) and protect from light.
Product Type LIS Enhancement Kit
Label or Dye NANOGOLD™, FluoroNanogold™, Silver, Colloidal Gold
Quantity 1 kit
Shipping Condition Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.