Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Naphthalene-2,3-Dicarboxaldehyde (NDA)

Catalog No. N1138
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1138 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1138 Supplier Invitrogen™ Supplier No. N1138
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Tagets amines

  • Aromatic dialdehyde naphthalene-2,3-dicarboxaldehyde (NDA) is essentially nonfluorescent until reacted with primary amine in presence of excess cyanide or thiol, to yield fluorescent isoindole with excitation/emission maxima approximately 419/493 nm

Anion Detection, Cell Analysis, Cell Viability, Proliferation and Function, Glutathione Detection, Ionic Homeostasis and Signaling, Nitro-Oxidative Stress, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature and protect from light.
Detection Method Fluorescence
Shipping Condition Room Temperature
Form Solid
Quantity 100 mg
Target Primary Amine
Type Naphthalene-2,3-Dicarboxaldehyde (NDA)
Label or Dye NDA (naphthalene-2,3-dicarboxaldehyde)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.