Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ovalbumin, Texas Red™ Conjugate

Catalog No. O23021
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O23021 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O23021 Supplier Invitrogen™ Supplier No. O23021
Add to Cart
Edge
Add to Cart

Labeled with bright, photostable and pH-insensitive Alexa Fluor 594 dye

The red fluorescent Texas Red ovalbumin conjugate is a protein with a relatively low molecular weight (∽45,000 daltons).

Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Conjugate Texas Red
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Classic Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Albumin Proteins
Product Line Texas Red
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.