Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rhodamine 110, bis-(p-Tosyl-L-Glycyl-L-Prolyl-L-Arginine Amide)

Catalog No. R22124
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R22124 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R22124 Supplier Invitrogen™ Supplier No. R22124
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This Molecular Probes™ bisamide derivative of rhodamine 110 (R110) is a sensitive and selective substrate for assaying proteases in solution or inside living cells.

This Molecular Probes™ bisamide derivative of rhodamine 110 (R110) is a sensitive and selective substrate for assaying proteases in solution or inside living cells. These fluorogenic substrates contain an amino acid or peptide covalently linked to each of R110's amino groups. Upon enzymatic cleavage, the nonfluorescent bisamide substrate is converted first to the fluorescent monoamide and then to R110, with a further increase in fluorescence. The substrate can be used to continuously measure enzyme activity in cell extracts and purified enzyme preparations using a fluorometer or fluorescence microplate reader, or can be used for intracellular protease assays with analysis by flow cytometry or fluorescence microscopy.<

• Has spectral properties similar to those of fluorescein, making it compatible with standard fluorescein filter sets
• Signal to background ratios are good because of large extinction coefficient of R110
• Unlike fluorescein, the fluorescence intensity of the rhodamine 110 reaction product is constant from pH 3–9
• Substrate name: p-tosyl-Gly-Pro-Arg)2-R110
• Useful to measure: Thrombin
• Ex/Em of R110 product: (498/521nm)
• Cell-Permeant

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Cell-Permeant
Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Form Solid
Quantity 2 mg
Substrate Caspase Substrate
Substrate Properties Peptide-Based Substrate
Target Enzyme Thrombin
For Use With (Equipment) Fluorometer, Microplate Reader
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.