Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Albumin from Bovine Serum (BSA), Texas Red™ conjugate
Description
- Applications include studying rate of endocytosis and exocytosis and following intracellular protein processing
Cell Analysis, Cell Tracing and Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Quantity | 5 mg |
| Conjugate | Texas Red |
| Species | Bovine |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Label Type | Classic Dyes |
| Shipping Condition | Room Temperature |
| Form | Lyophilized |
| Protein Subtype | Albumin Proteins |
| Product Line | Texas Red |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?