Learn More
DELTA ADSORBENTS 3A 4X8 Molecular Sieve Desiccant | 5lbs
Catalog No. NC1695064
Encompass
3A 4x8 Molecular Sieve is a multiple purpose, highly porous, high capacity alkali metal alumino-silicate in the spherical form. It is the potassium form of the Type A crystal structure with pore diameters of approximately 3Å. The 3Å pore opening will allow moisture to be adsorbed but will exclude unsaturated hydrocarbons and highly polar alcohols such as methanol and ethanol. It has high mass transfer efficiency and durability.
Catalog No. NC1695064 Supplier DELTA ADSORBENTS Supplier No. MS3A4805
3A 4x8 Molecular Sieve is a multiple purpose, highly porous, high capacity alkali metal alumino-silicate in the spherical form. It is the potassium form of the Type A crystal structure with pore diameters of approximately 3Å. The 3Å pore opening will allow moisture to be adsorbed but will exclude unsaturated hydrocarbons and highly polar alcohols such as methanol and ethanol. It has high mass transfer efficiency and durability.