Learn More
Sigma Aldrich Fine Chemicals Biosciences Molecular sieves 13X beads5KG
Catalog No. 502903939
Encompass
Molecular sieves 13X showed preferential adsorption of CO over nitrogen or hydrogen at all pressures up to 250psia.
Catalog No. 50-290-3939 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2086475KG
Molecular sieves 13X showed preferential adsorption of CO over nitrogen or hydrogen at all pressures up to 250psia.