Learn More
Sigma Aldrich Fine Chemicals Biosciences Molecular sieves 3 Abead5KG
Catalog No. 502898339
Encompass
The kinetics of the adsorption of water-vapor by employing molecular sieve 3A… has been studied in various temperature ranges (25-80C and water dew points from -69 to 17C).
Catalog No. 50-289-8339 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2085825KG
Add to Cart
Add to Cart
The kinetics of the adsorption of water-vapor by employing molecular sieve 3A… has been studied in various temperature ranges (25-80C and water dew points from -69 to 17C).