Learn More
Sigma Aldrich Fine Chemicals Biosciences Molecular sieves 4 A bead5KG
Catalog No. 502899831
Encompass
Regeneration or ActivationA saturated molecular sieve can be restored to its original capacity by regeneration the principle of which involves changing the conditions surrounding the adsorbent to correspond to a very low equilibrium capacity. In general the greater the difference between the equilibrium capacities of adsorption and regeneration the more rapid and complete the regeneration.The sieve may be regenerated in one of four ways1. Thermal reactivation -The maximum regeneration temperature for Silica is 300C.2. Pressure reactivation3. Passing an appropriate fluid through the gel bed at normal temperature and pressure.4. Displacement of adsorbates by passing a high concentration of molecules in a fluid through the bed.
Catalog No. 50-289-9831 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2085905KG
Regeneration or ActivationA saturated molecular sieve can be restored to its original capacity by regeneration the principle of which involves changing the conditions surrounding the adsorbent to correspond to a very low equilibrium capacity. In general the greater the difference between the equilibrium capacities of adsorption and regeneration the more rapid and complete the regeneration.The sieve may be regenerated in one of four ways1. Thermal reactivation -The maximum regeneration temperature for Silica is 300C.2. Pressure reactivation3. Passing an appropriate fluid through the gel bed at normal temperature and pressure.4. Displacement of adsorbates by passing a high concentration of molecules in a fluid through the bed.