Learn More
Sigma Aldrich Fine Chemicals Biosciences Molecular sieves, 5 Ã… powder, undried | 69912-79-4 | MFCD00131613 | 500G
Catalog No. NC2219344
Encompass
Molecular sieves, 5 Ã… powder, undried | 69912-79-4 | MFCD00131613 | 500G
This item is not returnable. View return policy
Catalog No. NC2219344 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 233676500G
Molecular sieves, 5 Ã… powder, undried | 69912-79-4 | MFCD00131613 | 500G