Learn More
Calcium hydride, ca. 93%, extra pure, 10-100 mm pieces, up to 10% powder
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 675.0°C |
| Color | Gray |
| Physical Form | Irregular Lumps/Pieces |
| Quantity | 2.5 kg |
| Assay Percent Range | 92% min. (Total Ca) |
| Linear Formula | CaH2 |
| Fieser | 01,105 |
| Merck Index | 15,1674 |
| Solubility Information | Solubility in water: reacts. Other solubilities: decomposes with lower alcohols and carboxylic acid, soluble only in salt melts |
| Formula Weight | 42.09 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Danger |
| missing translation for 'healthHazard2' | GHS H Statement In contact with water releases flammable gases which may ignite spontaneously. Causes skin irritation. Causes serious eye irritation. Reacts violently with water. |
| missing translation for 'healthHazard3' | GHS P Statement Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages. IF ON S |
| EINECSNumber | 232-189-2 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.