Learn More
Berry & Associates Inc Molecular Trap Bag, 2 g, highly activated molecular sieve, maintains water levels below 50 ppm in acetonitrile, prevents instrument clogging, packet
Catalog No. NC1812180
Encompass
Molecular Trap Bag, 2 g, highly activated molecular sieve, maintains water levels below 50 ppm in acetonitrile, prevents instrument clogging, packet
Catalog No. NC1812180 Supplier Berry & Associates Inc Supplier No. BG700061
Molecular Trap Bag, 2 g, highly activated molecular sieve, maintains water levels below 50 ppm in acetonitrile, prevents instrument clogging, packet