Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Molecular Weight Determination of Proteins
SDPSCI_ED

Catalog No. S11110 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
6 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S11110 6 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11110 Supplier Edvotek™ Supplier No. 110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

instructions, prestained LyphoProtein™ samples, gel loading solution, agarose, electrophoresis buffer, stain, SDS solution

Requires

  • Required Materials (not included):
  • Horizontal gel electrophoresis apparatus
  • DC power supply
  • Waterbath
  • Visualization system (white light)
  • Automatic micropipets with tips
  • Pipet pump
  • 500mL flasks
  • Distilled or deionized water
  • Methanol
  • Glacial acetic acid
  • Microwave
  • Hotplate or Bunsen burner

Introduce a simple method to determine protein subunit molecular weights using horizontal electrophoresis. Because the protein standards and 'unknowns' are prestained, the separation of proteins can be observed during electrophoresis.

Introduce a simple method to determine protein subunit molecular weights using horizontal electrophoresis. Because the protein standards and 'unknowns' are prestained, the separation of proteins can be observed during electrophoresis. Included in the experiment is EDVOTEK™ ́s formulation of protein grade agarose, which provides an alternative to the use of polyacrylamide gels.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Determine protein subunit molecular weights using horizontal electrophoresis
Includes Instructions, pre-stained LyphoProtein™ gel marker, pre-stained LyphoProtein™ samples, practice gel loading solution, Protein Agarose™ powder, Tris-Glycine-SDS powdered buffer, Protein InstaStain™, transfer pipets, and 1 mL pipet
Requires Horizontal electrophoresis apparatus, power supply, micropipette with tips, microwave oven or hot plate, 250 mL or 1 L flask, larger beaker, lab tape or markers, hot gloves, safety goggles and lab gloves, and distilled or deionized water. For Staining with Protein InstaStain™ (optional): methanol, glacial acetic acid, small plastic tray or large weigh boat, plastic wrap, white light box (recommended), rocking platform, and pipet pump or bulb
Time Range Gel Prep: 40 minutes; Electrophoresis: 60 minutes
Content And Storage Some components require freezer storage.
Class Size 6 Lab Groups
Product Type Electrophoresis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.