Learn More
Biochain Institute Inc 100 bp DNA Ladder Plus, ready-to-use, 250 ug, 0.5 ug per lane, 500 lanes/PK
SDP
Catalog No. 50181749
Encompass_Preferred
BioChain provides a series of molecular weight markers, including DNA markers, RNA markers, and protein markers, to accurately determine the size of your DNA, RNA, and protein on the separation gels.
Catalog No. 50-181-749 Supplier Biochain Institute Inc Supplier No. Z60300013
BioChain provides a series of molecular weight markers, including DNA markers, RNA markers, and protein markers, to accurately determine the size of your DNA, RNA, and protein on the separation gels.